Spermine tetrahydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

B1468-25G B1468-5G
TCB1468-25GEA 1405.95 CAD
TCB1468-25G TCB1468-005G
Spermine tetrahydrochloride ≥98.0% (by total nitrogen and titration analysis)
Spermine tetrahydrochloride

CAS Number: 306-67-2
MDL Number: MFCD00012914
Molecular Formula: C10H26N4
Molecular Weight: 348.18
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 315


Product Categories: Materials Chemistry, Monomers, Bioscience, Regenerative Medicine Research Reagents, Polyamines
Other Product Names / Synonyms: Spermine Tetrahydrochloride

Formula: C₁₀H₃₀Cl₄N₄
Melting Pt: 315 °C
MDL Number: MFCD00012914
CAS Number: 306-67-2
EINECS: 206-189-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df8aac133476ba7a56bcb69aba285d7e0618f6a5&vid=df8aac133476ba7a56bcb69aba285d7e0618f6a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR