Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)palladium(II) ≥90.0% (by titrimetric analysis)
Supplier: TCI America
B1437-1G
B1437-100MG
TCB1437-001GEA
270.18
CAD
TCB1437-001G
TCB1437-100MG
Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)palladium(II) ≥90.0% (by titrimetric analysis)
Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)palladium(II)
CAS Number: 72688-90-5
MDL Number: MFCD00143779
Molecular Formula: C38H72N2PdS10
Molecular Weight: 984.03
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Melting point (°C): 205
Formula:
C₃₈H₇₂N₂PdS₁₀ Melting Pt: 205 °C |
MDL Number:
MFCD00143779 CAS Number: 72688-90-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...