Ethyl-3,3-bis(methylthio)-2-cyanoacrylate ≥98.0%
Supplier: TCI America
Synonyms:
3,3-Bis(methylthio)-2-cyanoacrylic Acid Ethyl Ester, Ethyl 2-cyano-3,3-bis(methylthio)acrylate
B1411-500G
B1411-25G
TCB1411-500GEA
1510.8
CAD
TCB1411-500G
TCB1411-025G
Ethyl-3,3-bis(methylthio)-2-cyanoacrylate ≥98.0%
Ethyl-3,3-bis(methylthio)-2-cyanoacrylate
CAS Number: 17823-58-4
MDL Number: MFCD00143027
Molecular Formula: C8H11NO2S2
Molecular Weight: 217.30
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 58
Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: 3,3-Bis(methylthio)-2-cyanoacrylic Acid Ethyl Ester
Formula:
C₈H₁₁NO₂S₂ MW: 217.30 g/mol Melting Pt: 58 °C |
MDL Number:
MFCD00143027 CAS Number: 17823-58-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...