4,4'-(Hexafluoroisopropylidene)dianiline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B1400-25G
B1400-5G
B1400-1G
TCB1400-25GEA
1303.5
CAD
TCB1400-25G
TCB1400-005G
76219-336
4,4'-(Hexafluoroisopropylidene)dianiline ≥98.0% (by GC, titration analysis)
4,4'-(Hexafluoroisopropylidene)dianiline
CAS Number: 1095-78-9
MDL Number: MFCD00039146
Molecular Formula: C15H12F6N2
Molecular Weight: 334.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 198
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Bisphenol AF type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-(Hexafluoroisopropylidene)dianiline
Formula:
C₁₅H₁₂F₆N₂ Melting Pt: 198 °C |
MDL Number:
MFCD00039146 CAS Number: 1095-78-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...