2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B1398-5G B1398-25G
TCB1398-005GEA 91.64 CAD
TCB1398-005G TCB1398-025G
2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane ≥98.0% (by HPLC, titration analysis)
2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane

CAS Number: 69563-88-8
MDL Number: MFCD00015723
Molecular Formula: C27H20F6N2O2
Molecular Weight: 518.46
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 163


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Bisphenol AF type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-(Hexafluoroisopropylidene)bis[(4-aminophenoxy)benzene]

Formula: C₂₇H₂₀F₆N₂O₂
Melting Pt: 163 °C
MDL Number: MFCD00015723
CAS Number: 69563-88-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a94d109f5f3327de64f59cdc21fa46211772bf52&vid=a94d109f5f3327de64f59cdc21fa46211772bf52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR