8-Amino-2-methylquinoline ≥98.0%
Supplier: TCI America
Synonyms:
2-Methylquinolin-8-amine, 8-Aminoquinaldine
A0416-5G
A0416-1G
TCA0416-5GEA
277.76
CAD
TCA0416-5G
TCA0416-001G
8-Amino-2-methylquinoline ≥98.0%
8-Amino-2-methylquinoline
CAS Number: 18978-78-4
MDL Number: MFCD00023998
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 56
Other Product Names / Synonyms: 8-Aminoquinaldine
Formula:
C₁₀H₁₀N₂ Melting Pt: 56 °C |
MDL Number:
MFCD00023998 CAS Number: 18978-78-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...