4-tert-Butyl-3-methoxy-2,6-dinitrotoluene ≥99.0%

Supplier: TCI America

B1379-25G B1379-500G
TCB1379-25GEA 98.86 CAD
TCB1379-25G TCB1379-500G
4-tert-Butyl-3-methoxy-2,6-dinitrotoluene ≥99.0%
4-tert-Butyl-3-methoxy-2,6-dinitrotoluene

CAS Number: 83-66-9
MDL Number: MFCD00024268
Molecular Formula: C12H16N2O5
Molecular Weight: 268.27
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 86


Other Product Names / Synonyms: 6-tert-Butyl-3-methyl-2,4-dinitroanisole

Formula: C₁₂H₁₆N₂O₅
Boiling Pt: 185 °C
Melting Pt: 86 °C
MDL Number: MFCD00024268
CAS Number: 83-66-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceb7a5ac4d416520b5debaa6fcf28a6ec078c085&vid=ceb7a5ac4d416520b5debaa6fcf28a6ec078c085&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR