1,3-Bis(3-aminophenoxy)benzene ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B1370-25G B1370-5G
TCB1370-25GEA 380.78 CAD
TCB1370-25G TCB1370-005G
1,3-Bis(3-aminophenoxy)benzene ≥98.0% (by HPLC, titration analysis)
1,3-Bis(3-aminophenoxy)benzene

CAS Number: 10526-07-5
MDL Number: MFCD00043718
Molecular Formula: C18H16N2O2
Molecular Weight: 292.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 108
Flash Point (°C): 102


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Diphenyl Ethers (for High-Performance Polymer Research)
Other Product Names / Synonyms: Resorcinol Bis(3-aminophenyl) Ether

Formula: C₁₈H₁₆N₂O₂
MW: 292.3318 g/mol
Melting Pt: 108 °C
Flash Pt: 102 °C
MDL Number: MFCD00043718
CAS Number: 10526-07-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37d78a4da8a37a9166cb78f811cf0af2dc6b48ac&vid=37d78a4da8a37a9166cb78f811cf0af2dc6b48ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR