4-(Benzyloxy)benzonitrile ≥98.0%

Supplier: TCI America

B1365-25G B1365-5G
TCB1365-025GEA 138.88 CAD
TCB1365-025G TCB1365-5G
4-(Benzyloxy)benzonitrile ≥98.0%
4-(Benzyloxy)benzonitrile

CAS Number: 52805-36-4
MDL Number: MFCD00079701
Molecular Formula: C14H11NO
Molecular Weight: 209.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 94

Formula: C₁₄H₁₁NO
Melting Pt: 94 °C
MDL Number: MFCD00079701
CAS Number: 52805-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebaa4cdb0a1a11f5d03b19f67cb3594f3a83d036&vid=ebaa4cdb0a1a11f5d03b19f67cb3594f3a83d036&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR