N-Benzyliminodiacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1363-25G B1363-5G
TCB1363-025GEA 545.1 CAD
TCB1363-025G TCB1363-005G
N-Benzyliminodiacetic acid ≥98.0% (by titrimetric analysis)
N-Benzyliminodiacetic acid

CAS Number: 3987-53-9
MDL Number: MFCD00004285
Molecular Formula: C11H13NO4
Molecular Weight: 223.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 201

Formula: C₁₁H₁₃NO₄
Melting Pt: 201 °C
MDL Number: MFCD00004285
CAS Number: 3987-53-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1347f5226d4a488b26abac5f0ffedae0cdb75af7&vid=1347f5226d4a488b26abac5f0ffedae0cdb75af7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR