3-Boc-aminopyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(BOC-amino)pyrrolidine, 2-Methyl-2-propanyl 3-pyrrolidinylcarbamate, 1,1-Dimethylethyl3-pyrrolidinylcarbamate, 3-Boc-Aminopyrrolidine

B1358-5G B1358-25G
TCB1358-005GEA 74.26 CAD
TCB1358-005G TCB1358-25G
3-Boc-aminopyrrolidine ≥98.0% (by GC, titration analysis)
3-Boc-aminopyrrolidine

CAS Number: 99724-19-3
MDL Number: MFCD00059040
Molecular Formula: C9H18N2O2
Molecular Weight: 186.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 54


Other Product Names / Synonyms: 3-(Boc-amino)pyrrolidine

Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Melting Pt: 54 °C
MDL Number: MFCD00059040
CAS Number: 99724-19-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41aec663824acd926ce6cc0e1cf3019a464279da&vid=41aec663824acd926ce6cc0e1cf3019a464279da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR