(4-(Bromomethyl)phenyl)acetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B1351-25G
B1351-5G
TCB1351-25GEA
243.32
CAD
TCB1351-25G
TCB1351-005G
(4-(Bromomethyl)phenyl)acetic acid ≥98.0% (by titrimetric analysis)
(4-(Bromomethyl)phenyl)acetic acid
CAS Number: 13737-36-5
MDL Number: MFCD00010632
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 180
Other Product Names / Synonyms: 4-(Carboxymethyl)benzyl Bromide
Formula:
(C₆H₄(CH₂Br))CH₂COOH Melting Pt: 180 °C |
MDL Number:
MFCD00010632 CAS Number: 13737-36-5 UN: 3261 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...