(2S,3S)-(+)-2,3-Butanediol ≥97.0%

Supplier: TCI America

Synonyms: (2S,3S)-Butane-2,3-diol, (S,S)-(+)-2,3-Butanediol, (2S,3S)-(+)-2,3-Butylene glykol, (S,S)-(+)-2,3-Dihydroxy butane, (2S,3S)-(+)-2,3-Dihydroxy butane, (S,S)-(+)-2,3-Butylene glykol, L(+)-2,3-Butanediol

B1343-1G B1343-100MG
TCB1343-1GEA 195.92 CAD
TCB1343-1G TCB1343-100MG
(2S,3S)-(+)-2,3-Butanediol ≥97.0%
(2S,3S)-(+)-2,3-Butanediol

CAS Number: 19132-06-0
MDL Number: MFCD00063648
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 182
Flash Point (°C): 85
Freezing point (°C): 17
Specific Gravity (20/20): 0.99
Specific rotation [a]20/D: 13.5 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (S,S)-(+)-2,3-Butylene Glycol, (S,S)-(+)-2,3-Dihydroxybutane

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 182 °C
Density: 0.99 (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00063648
CAS Number: 19132-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6fa87e382cbf2fbe97eeacf820dda5dd6578f65&vid=f6fa87e382cbf2fbe97eeacf820dda5dd6578f65&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR