4-tert-Butylphthalic anhydride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B1337-5G
B1337-25G
TCB1337-5GEA
173.8
CAD
TCB1337-5G
TCB1337-025G
4-tert-Butylphthalic anhydride ≥98.0% (by GC, titration analysis)
4-tert-Butylphthalic anhydride
CAS Number: 32703-79-0
MDL Number: MFCD00060127
Molecular Formula: C12H12O3
Molecular Weight: 204.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 78
Formula:
C₁₂H₁₂O₃ Boiling Pt: 136 °C Melting Pt: 78 °C |
MDL Number:
MFCD00060127 CAS Number: 32703-79-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...