[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1326-25G B1326-100G
TCB1326-025GEA 34.76 CAD
TCB1326-025G TCB1326-100G
[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone ≥98.0% (by titrimetric analysis)
[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone

CAS Number: 2420-87-3
MDL Number: MFCD00039140
Molecular Formula: C16H6O6
Molecular Weight: 294.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 303


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: Diphenyl-3,3',4,4'-tetracarboxylic Dianhydride

Formula: C16H6O6
Melting Pt: 303 °C
MDL Number: MFCD00039140
CAS Number: 2420-87-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=459fc3a72d241bc35b56a2fa1412c580255314d2&vid=459fc3a72d241bc35b56a2fa1412c580255314d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR