3-Amino-4-hydroxybenzenesulfonic acid hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0399-25G A0399-500G
TCA0399-025GEA 64.78 CAD
TCA0399-025G TCA0399-500G
3-Amino-4-hydroxybenzenesulfonic acid hydrate ≥98.0% (by titrimetric analysis)
3-Amino-4-hydroxybenzenesulfonic acid hydrate

CAS Number: 98-37-3
MDL Number: MFCD00007696
Molecular Formula: C6H7NO4S
Molecular Weight: 189.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 220


Other Product Names / Synonyms: 2-Aminophenol-4-sulfonic Acid, 4-Hydroxymetanilic Acid

Formula: C₆H₇NO₄S
Melting Pt: 220 °C
MDL Number: MFCD00007696
CAS Number: 98-37-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b95cb9d45d33dc819ef55ae427fe361aed50450&vid=7b95cb9d45d33dc819ef55ae427fe361aed50450&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR