Phenacyl-4-(bromomethyl)phenylacetate ≥97.0%

Supplier: TCI America

Synonyms: 4-(Bromomethyl)phenylacetic Acid Phenacyl Ester

B1315-1G B1315-5G
TCB1315-1GEA 83.74 CAD
TCB1315-1G TCB1315-5G
Phenacyl-4-(bromomethyl)phenylacetate ≥97.0%
Phenacyl-4-(bromomethyl)phenylacetate

CAS Number: 66270-97-1
MDL Number: MFCD00009632
Molecular Formula: C17H15BrO3
Molecular Weight: 347.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 88


Other Product Names / Synonyms: 4-(Bromomethyl)phenylacetic Acid Phenacyl Ester

Formula: C₁₇H₁₅BrO₃
MW: 347.21 g/mol
Melting Pt: 88 °C
MDL Number: MFCD00009632
CAS Number: 66270-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ae95265981221af5065eaefb81ca467d3111d&vid=9a6ae95265981221af5065eaefb81ca467d3111d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR