4-Bromo-4'-propylbiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromo-4'-propyl-1,1'-biphenyl

B1306-25G B1306-5G
TCB1306-25GEA 338.91 CAD
TCB1306-25G TCB1306-005G
4-Bromo-4'-propylbiphenyl ≥98.0%
4-Bromo-4'-propylbiphenyl

CAS Number: 58743-81-0
MDL Number: MFCD00060105
Molecular Formula: C15H15Br
Molecular Weight: 275.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 108

Formula: C₁₅H₁₅Br
MW: 275.1800 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00060105
CAS Number: 58743-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=325b1bfb74c55f2d0b437adfe3932c5760912ee8&vid=325b1bfb74c55f2d0b437adfe3932c5760912ee8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR