Victoria pure blue bo

Supplier: TCI America

B1301-250G B1301-25G
TCB1301-250GEA 657.28 CAD
TCB1301-250G TCB1301-025G
Victoria pure blue bo
Victoria Pure Blue Bo

CAS Number: 2390-60-5
MDL Number: MFCD00011877
Molecular Formula: C33H40ClN3
Molecular Weight: 514.15
Form: Crystal
Color: Yellow


Other Product Names / Synonyms: Victoria Pure Blue BO

Formula: C₃₃H₄₀ClN₃
MDL Number: MFCD00011877
CAS Number: 2390-60-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4badfcc081722b2a1ca9fa33484d4a05fbb8d39&vid=c4badfcc081722b2a1ca9fa33484d4a05fbb8d39&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR