trans-4-tert-Butylcyclohexanol ≥95.0%
Supplier: TCI America
B1271-1G
B1271-5G
TCB1271-001GEA
369.72
CAD
TCB1271-001G
TCB1271-005G
trans-4-tert-Butylcyclohexanol ≥95.0%
trans-4-tert-Butylcyclohexanol
CAS Number: 21862-63-5
MDL Number: MFCD00064952
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 80
Formula:
C₁₀H₂₀O Melting Pt: 80 °C |
MDL Number:
MFCD00064952 CAS Number: 21862-63-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...