cis-4-tert-Butylcyclohexanol ≥98.0%
Supplier: TCI America
B1270-1G
B1270-5G
TCB1270-001GEA
146.31
CAD
TCB1270-001G
TCB1270-5G
cis-4-tert-Butylcyclohexanol ≥98.0%
cis-4-tert-Butylcyclohexanol
CAS Number: 937-05-3
MDL Number: MFCD00070476
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 83
Formula:
C₁₀H₂₀O Melting Pt: 83 °C |
MDL Number:
MFCD00070476 CAS Number: 937-05-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...