2-Amino-4-phenylphenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A0397-25G A0397-1G A0397-5G
TCA0397-25GEA 532.3 CAD
TCA0397-25G TCA0397-001G TCA0397-005G
2-Amino-4-phenylphenol ≥98.0% (by GC, titration analysis)
2-Amino-4-phenylphenol

CAS Number: 1134-36-7
MDL Number: MFCD00059187
Molecular Formula: C12H11NO
Molecular Weight: 185.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 207


Other Product Names / Synonyms: 3-Aminobiphenyl-4-ol, 3-Amino-4-hydroxybiphenyl

Formula: C₁₂H₁₁NO
Melting Pt: 207 °C
MDL Number: MFCD00059187
CAS Number: 1134-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9980011df2f29025c40bad77e66417b07a227a32&vid=9980011df2f29025c40bad77e66417b07a227a32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR