Benzyl isopropenyl ether ≥97.0%

Supplier: TCI America

Synonyms: 2-Benzyloxy-1-propene, Isopropenyl Benzyl Ether

B1257-5ML B1257-1ML
TCB1257-5MLEA 756.82 CAD
TCB1257-5ML TCB1257-001ML
Benzyl isopropenyl ether ≥97.0%
Benzyl isopropenyl ether

[Hydroxyl-Protecting Agent]
CAS Number: 32783-20-3
MDL Number: MFCD00059913
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 192
Specific Gravity (20/20): 0.96
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: 2-Benzyloxy-1-propene, Isopropenyl Benzyl Ether

Formula: C₁₀H₁₂O
MW: 148.21 g/mol
Boiling Pt: 192 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00059913
CAS Number: 32783-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=981d49529e3133dcc2b068e6a0854c609bc50e1b&vid=981d49529e3133dcc2b068e6a0854c609bc50e1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR