1-Butyl-2-pyrrolidone ≥98.0%

Supplier: TCI America

B1254-25ML B1254-5ML
TCB1254-025MLEA 340.74 CAD
TCB1254-025ML 76219-334
1-Butyl-2-pyrrolidone ≥98.0%
1-Butyl-2-pyrrolidone

CAS Number: 3470-98-2
MDL Number: MFCD00020876
Molecular Formula: C8H15NO
Molecular Weight: 141.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: N-Butyl-2-azacyclopentanone

Formula: C₈H₁₅NO
Boiling Pt: 137 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00020876
CAS Number: 3470-98-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fbf635588eac0febf6f72f7dc14b1655a56e682&vid=1fbf635588eac0febf6f72f7dc14b1655a56e682&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR