Benzo-12-crown-4 (Benzo-1,4,7,10-tetraoxacyclododecane) ≥98.0%
Supplier: TCI America
Synonyms:
Benzo-12-crown 4-Ether, 2,3-Benzo-1,4,7,10-tetraoxadodec-2-ene, Benzo-12-crown-4
B1249-1G
B1249-5G
TCB1249-1GEA
195.92
CAD
TCB1249-1G
TCB1249-005G
Benzo-12-crown-4 (Benzo-1,4,7,10-tetraoxacyclododecane) ≥98.0%
Benzo-12-crown-4 (Benzo-1,4,7,10-tetraoxacyclododecane)
CAS Number: 14174-08-4
MDL Number: MFCD00059934
Molecular Formula: C12H16O4
Molecular Weight: 224.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 46
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-Benzo-1,4,7,10-tetraoxadodec-2-ene
Formula:
C₁₂H₁₆O₄ MW: 224.26 g/mol Melting Pt: 46 °C |
MDL Number:
MFCD00059934 CAS Number: 14174-08-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...