4-Hydroxyanilinium chloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 4-Aminophenol hydrochloride 4-Hydroxyaniline hydrochloride, p-Aminophenol hydrochloride, 4-Aminophenol hydrochloride

A0392-25G A0392-100G
TCA0392-025GEA 104.28 CAD
TCA0392-025G TCA0392-100G
4-Hydroxyanilinium chloride ≥98.0% (by HPLC, total nitrogen)
4-Hydroxyanilinium chloride

CAS Number: 51-78-5
MDL Number: MFCD00012996
Molecular Formula: C6H7NO
Molecular Weight: 145.59
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 306

Formula: C₆H₈ClNO
MW: 145.59 g/mol
Melting Pt: 306 °C
MDL Number: MFCD00012996
CAS Number: 51-78-5
EINECS: 200-122-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a4703e0b8c6170852bd192d2ea7e6e9c3e1a456&vid=9a4703e0b8c6170852bd192d2ea7e6e9c3e1a456&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR