1,8-Bis(2-pyridyl)-3,6-dithiaoctane ≥98.0%
Supplier: TCI America
Synonyms:
S,S'-Bis[2-(2-pyridyl)ethyl]-1,2-ethanedithiol, 2,2'-(3,6-Dithiaoctamethylene)pyridine
B1229-5G
B1229-1G
TCB1229-5GEA
1281.38
CAD
TCB1229-5G
TCB1229-001G
1,8-Bis(2-pyridyl)-3,6-dithiaoctane ≥98.0%
1,8-Bis(2-pyridyl)-3,6-dithiaoctane
CAS Number: 64691-70-9
MDL Number: MFCD00006366
Molecular Formula: C16H20N2S2
Molecular Weight: 304.47
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 109
Other Product Names / Synonyms: S,S'-Bis[2-(2-pyridyl)ethyl]-1,2-ethanedithiol, 2,2'-(3,6-Dithiaoctamethylene)pyridine
Formula:
C₁₆H₂₀N₂S₂ Melting Pt: 51 °C Flash Pt: 109 °C |
MDL Number:
MFCD00006366 CAS Number: 64691-70-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...