(±)-α-Bromo-γ-butyrolactone ≥95.0%

Supplier: TCI America

B1228-25G B1228-500G
TCB1228-025GEA 67.94 CAD
TCB1228-025G TCB1228-500G
(±)-α-Bromo-γ-butyrolactone ≥95.0%
(±)-α-Bromo-γ-butyrolactone

CAS Number: 5061-21-2
MDL Number: MFCD00005387
Molecular Formula: C4H5BrO2
Molecular Weight: 164.99
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 110
Specific Gravity (20/20): 1.80

Formula: C₄H₅BrO₂
Boiling Pt: 140 °C
Density: 1.80 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005387
CAS Number: 5061-21-2
EINECS: 225-764-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cd45c79b295b8d72243e2f063cea52ad79779ad&vid=7cd45c79b295b8d72243e2f063cea52ad79779ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR