4-Butylbenzaldehyde diethyl acetal ≥97.0%

Supplier: TCI America

B1222-5ML B1222-25ML
TCB1222-5MLEA 142.2 CAD
TCB1222-5ML TCB1222-025ML
4-Butylbenzaldehyde diethyl acetal ≥97.0%
4-Butylbenzaldehyde diethyl acetal

CAS Number: 83803-80-9
MDL Number: MFCD00009466
Molecular Formula: C15H24O2
Molecular Weight: 236.36
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 0.93

Formula: C₁₅H₂₄O₂
Density: 0.93 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00009466
CAS Number: 83803-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=050a4aea4d904122181b94279729bef0ced25d5b&vid=050a4aea4d904122181b94279729bef0ced25d5b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR