Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc complex ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
Synonyms:
BTBA-BDTD Zn Complex
B1221-25G
B1221-5G
TCB1221-025GEA
1166.04
CAD
TCB1221-025G
TCB1221-005G
Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc complex ≥98.0% (by total nitrogen and titration analysis)
Bis(tetrabutylammonium) bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc complex
CAS Number: 68449-38-7
MDL Number: MFCD00060267
Molecular Formula: C38H72N2S10Zn
Molecular Weight: 942.99
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 174
Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: BTBA-BDTD Zn Complex
Formula:
C₃₈H₇₂N₂S₁₀Zn MW: 942.99 g/mol Melting Pt: 174 °C |
MDL Number:
MFCD00060267 CAS Number: 68449-38-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...