4,4'-Bis(dimethylamino)benzil ≥99.0% (by titrimetric analysis)
Supplier: TCI America
B1212-5G
B1212-25G
TCB1212-005GEA
236.37
CAD
TCB1212-005G
TCB1212-25G
4,4'-Bis(dimethylamino)benzil ≥99.0% (by titrimetric analysis)
4,4'-Bis(dimethylamino)benzil
CAS Number: 17078-27-2
MDL Number: MFCD00051389
Molecular Formula: C18H20N2O2
Molecular Weight: 296.37
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 203
Other Product Names / Synonyms: 1,2-Bis(4-dimethylaminophenyl)-1,2-ethanedione
Formula:
C₁₈H₂₀N₂O₂ Melting Pt: 203 °C |
MDL Number:
MFCD00051389 CAS Number: 17078-27-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...