4,5-Bis(benzoylthio)-1,3-dithiole-2-thione ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B1199-1G
B1199-5G
TCB1199-001GEA
48.98
CAD
TCB1199-001G
TCB1199-005G
4,5-Bis(benzoylthio)-1,3-dithiole-2-thione ≥98.0% (by titrimetric analysis)
4,5-Bis(benzoylthio)-1,3-dithiole-2-thione
CAS Number: 68494-08-6
MDL Number: MFCD00059894
Molecular Formula: C17H10O2S5
Molecular Weight: 406.57
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 142
Formula:
C₁₇H₁₀O₂S₅ MW: 406.57 g/mol Melting Pt: 142 °C |
MDL Number:
MFCD00059894 CAS Number: 68494-08-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...