(±)-2-Aminooctanoic acid ≥98.0%
Supplier: TCI America
Synonyms:
DL-α-Aminocaprylic acid, 2-Aminooctanoic acid
A0382-5G
A0382-25G
TCA0382-5GEA
139.04
CAD
TCA0382-5G
TCA0382-025G
(±)-2-Aminooctanoic acid ≥98.0%
(±)-2-Aminooctanoic acid
CAS Number: 644-90-6
MDL Number: MFCD00008102
Molecular Formula: C8H17NO2
Molecular Weight: 159.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₈H₁₇NO₂ |
MDL Number:
MFCD00008102 CAS Number: 644-90-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...