Biphenyl-4,4'-dicarboxylic acid ≥97.0% for synthesis

Supplier: TCI America

Synonyms: 4,4'-Biphenyldicarboxylic acid, [1,1'-Biphenyl]-4,4'-dicarboxylic acid

B1191-5G B1191-25G
TCB1191-005GEA 126.4 CAD
TCB1191-005G TCB1191-025G
Biphenyl-4,4'-dicarboxylic acid ≥97.0% for synthesis
Biphenyl-4,4'-dicarboxylic acid

CAS Number: 787-70-2
MDL Number: MFCD00002554
Molecular Formula: C14H10O4
Molecular Weight: 242.23
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Monomers, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Bibenzoic Acid, 4,4'-Dibenzoic Acid, 4,4'-Diphenyldicarboxylic Acid

Formula: C₁₄H₁₀O₄
MDL Number: MFCD00002554
CAS Number: 787-70-2
EINECS: 212-328-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82f79d0147604db7fcc11fcf00789f72b6f54706&vid=82f79d0147604db7fcc11fcf00789f72b6f54706&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR