7-Bromo-5-chloro-8-hydroxyquinoline ≥98.0%

Supplier: TCI America

Synonyms: 7-Bromo-5-chloroquinolin-8-ol

B1190-5G B1190-1G
TCB1190-005GEA 575.75 CAD
TCB1190-005G TCB1190-1G
7-Bromo-5-chloro-8-hydroxyquinoline ≥98.0%
7-Bromo-5-chloro-8-hydroxyquinoline

CAS Number: 7640-33-7
MDL Number: MFCD00044601
Molecular Formula: C9H5BrClNO
Molecular Weight: 258.50
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 181

Formula: C₉H₅BrClNO
Melting Pt: 181 °C
MDL Number: MFCD00044601
CAS Number: 7640-33-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=786f62b90e64f1a14b136753b410f4641557760b&vid=786f62b90e64f1a14b136753b410f4641557760b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR