1-Benzyl-1,4-dihydronicotinamide ≥95.0%

Supplier: TCI America

B1156-1G B1156-25G B1156-5G
TCB1156-001GEA 104.28 CAD
TCB1156-001G TCB1156-025G TCB1156-5G
1-Benzyl-1,4-dihydronicotinamide ≥95.0%
1-Benzyl-1,4-dihydronicotinamide

CAS Number: 952-92-1
MDL Number: MFCD00059769
Molecular Formula: C13H14N2O
Molecular Weight: 214.27
Purity/Analysis Method: >95.0% (HPLC,N)
Form: Crystal
Melting point (°C): 121
Storage Temperature: 0-10°C

Formula: C₁₃H₁₄N₂O
Melting Pt: 121 °C
MDL Number: MFCD00059769
CAS Number: 952-92-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=848fff73117595c086ecb51604ed47f80d1dfe32&vid=848fff73117595c086ecb51604ed47f80d1dfe32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR