N-Benzoyl-L-tyrosine ethyl ester ≥98.0%
Supplier: TCI America
Synonyms:
BTEE, Bz-Tyr-OEt
B1140-5G
B1140-1G
TCB1140-005GEA
95.91
CAD
TCB1140-005G
TCB1140-001G
N-Benzoyl-L-tyrosine ethyl ester ≥98.0%
N-Benzoyl-L-tyrosine ethyl ester
CAS Number: 3483-82-7
MDL Number: MFCD00002388
Molecular Formula: C18H19NO4
Molecular Weight: 313.35
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 122
Specific rotation [a]20/D: -29 deg (C=1, EtOH)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: BTEE, Bz-Tyr-OEt
Formula:
C₁₈H₁₉NO₄ Melting Pt: 122 °C |
MDL Number:
MFCD00002388 CAS Number: 3483-82-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...