trans-4-Butylcyclohexanecarboxylic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

B1136-5G B1136-25G
TCB1136-005GEA 121.66 CAD
TCB1136-005G TCB1136-025G
trans-4-Butylcyclohexanecarboxylic acid ≥99.0% (by GC, titration analysis)
trans-4-Butylcyclohexanecarboxylic acid

CAS Number: 38289-28-0
MDL Number: MFCD00059560
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 37
Storage Temperature: 0-10°C

Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Melting Pt: 37 °C
MDL Number: MFCD00059560
CAS Number: 38289-28-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b33138a6c5ec1bef8929fac84f4a142781bbace5&vid=b33138a6c5ec1bef8929fac84f4a142781bbace5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR