2-(5-Bromo-2-pyridylazo)-5-dimethylaminophenol ≥93.0%

Supplier: TCI America

Synonyms: 2-(4-Dimethylamino-2-hydroxyphenylazo)-5-bromopyridine

B1082-1G B1082-100MG
TCB1082-001GEA 2355.78 CAD
TCB1082-001G TCB1082-100MG
2-(5-Bromo-2-pyridylazo)-5-dimethylaminophenol ≥93.0%
2-(5-Bromo-2-pyridylazo)-5-dimethylaminophenol

[for Colorimetric Determination of Cd]
CAS Number: 50783-82-9
MDL Number: MFCD00059779
Molecular Formula: C13H13BrN4O
Molecular Weight: 321.18
Purity/Analysis Method: >93.0% (HPLC)
Form: Crystal
Melting point (°C): 217


Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 2-(4-Dimethylamino-2-hydroxyphenylazo)-5-bromopyridine

Formula: C₁₃H₁₃BrN₄O
Melting Pt: 217 °C
MDL Number: MFCD00059779
CAS Number: 50783-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ad763acebc30accaa7193074c1de8178d084ff7&vid=5ad763acebc30accaa7193074c1de8178d084ff7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR