2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol ≥98.0%

Supplier: TCI America

Synonyms: 2-((5-Bromopyridin-2-yl)diazenyl)-5-(diethylamino)phenol, 2-(4-Diethylamino-2-hydroxyphenylazo)-5-bromopyridine

B1081-1G B1081-100MG
TCB1081-001GEA 297.51 CAD
TCB1081-001G TCB1081-100MG
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol ≥98.0%
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol

CAS Number: 14337-53-2
MDL Number: MFCD00006255
Molecular Formula: C15H17BrN4O
Molecular Weight: 349.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 157


Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Other Product Names / Synonyms: 2-(4-Diethylamino-2-hydroxyphenylazo)-5-bromopyridine

Formula: C₁₅H₁₇BrN₄O
Melting Pt: 157 °C
MDL Number: MFCD00006255
CAS Number: 14337-53-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2791abff7e657e7025f567d5a620e008f55131b&vid=c2791abff7e657e7025f567d5a620e008f55131b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR