2,2'-Bicinchoninic acid dipotassium salt hydrate ≥98.0% (by HPLC)
Supplier: TCI America
[for Determination of Cu]
CAS Number: 63451-34-3
MDL Number: MFCD00014664
Molecular Formula: C20H12N2O4
Molecular Weight: 420.51
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Ligands for Functional Metal Complexes, Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: 2,2'-Biquinoline-4,4'-dicarboxylic Acid Dipotassium Salt, 4,4'-Dicarboxy-2,2'-biquinoline Dipotassium Salt
Formula:
C₂₀H₁₂N₂O₄ |
MDL Number:
MFCD00014664 CAS Number: 63451-34-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...