1,4-Butanediolbis(3-aminocrotonate) ≥96.0%

Supplier: TCI America

Synonyms: 1,4-Butylene Bis(3-aminocrotonate)

B1069-25G B1069-500G
TCB1069-025GEA 36.34 CAD
TCB1069-025G TCB1069-500G
1,4-Butanediolbis(3-aminocrotonate) ≥96.0%
1,4-Butanediolbis(3-aminocrotonate)

CAS Number: 14205-47-1
MDL Number: MFCD00059862
Molecular Formula: C12H20N2O4
Molecular Weight: 256.30
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 148


Other Product Names / Synonyms: 1,4-Butylene Bis(3-aminocrotonate)

Formula: C₁₂H₂₀N₂O₄
MW: 256.30 g/mol
Melting Pt: 148 °C
MDL Number: MFCD00059862
CAS Number: 14205-47-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=641800c76036b8e79185ce06fbc28dcdd4c987c8&vid=641800c76036b8e79185ce06fbc28dcdd4c987c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR