3-Bromophthalide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1068-25G B1068-5G
TCB1068-025GEA 229.1 CAD
TCB1068-025G TCB1068-5G
3-Bromophthalide ≥98.0% (by titrimetric analysis)
3-Bromophthalide

CAS Number: 6940-49-4
MDL Number: MFCD00040520
Molecular Formula: C8H5BrO2
Molecular Weight: 213.03
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 138
Melting point (°C): 86
Flash Point (°C): 100


Other Product Names / Synonyms: 3-Bromo-1(3H)-isobenzofuranone

Formula: C₈H₅BrO₂
Boiling Pt: 138 °C
Melting Pt: 86 °C
Flash Pt: 100 °C
MDL Number: MFCD00040520
CAS Number: 6940-49-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efe2866da3f20d6c3f3535b9152e71eb50a87291&vid=efe2866da3f20d6c3f3535b9152e71eb50a87291&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR