1,3-Butanediol dimethacrylate ≥98.0% stabilised for synthesis

Supplier: TCI America

Synonyms: 1,3-Butylene glycol dimethacrylate

B1065-25ML B1065-500ML
TCB1065-025MLEA 43.42 CAD
TCB1065-025ML TCB1065-500ML
1,3-Butanediol dimethacrylate ≥98.0% stabilised for synthesis
1,3-Butanediol dimethacrylate

(stabilized with MEHQ)
CAS Number: 1189-08-8
MDL Number: MFCD00014930
Molecular Formula: C12H18O4
Molecular Weight: 226.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 290
Flash Point (°C): 130
Specific Gravity (20/20): 1.01


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Butylene Glycol Dimethacrylate

Formula: C₁₂H₁₈O₄
MW: 226.27 g/mol
Boiling Pt: 290 °C
Density: 1.01 (20 °C)
Flash Pt: 130 °C
MDL Number: MFCD00014930
CAS Number: 1189-08-8
EINECS: 214-711-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb9b87a3305fae8b47dbff9cfcdff7a1690e043e&vid=eb9b87a3305fae8b47dbff9cfcdff7a1690e043e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR