4,4'-(1-Methylpropylidene)diphenol ≥98.0%

Supplier: TCI America

Synonyms: Bisphenol B, 2,2-Bis(4-hydroxyphenyl)butane, 4,4'-(Butane-2,2-diyl)diphenol, 4,4'-Butane-2,2-diyldiphenol

B1050-5G B1050-25G
TCB1050-005GEA 229.1 CAD
TCB1050-005G TCB1050-025G
4,4'-(1-Methylpropylidene)diphenol ≥98.0%
4,4'-(1-Methylpropylidene)diphenol

CAS Number: 77-40-7
MDL Number: MFCD00020179
Molecular Formula: C16H18O2
Molecular Weight: 242.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 126


Product Categories: Materials Chemistry, Monomers, Bisphenol A type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bisphenol B

Formula: C₁₆H₁₈O₂
MW: 242.32 g/mol
Boiling Pt: 412
Melting Pt: 126 °C
MDL Number: MFCD00020179
CAS Number: 77-40-7
EINECS: 201-025-1
Merck Index: 12,01339
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57498c0dc26a93298a349a2531da13ca1fbf4df8&vid=57498c0dc26a93298a349a2531da13ca1fbf4df8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR