6-Amino-1-naphthol-3-sulfonic acid hydrate ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 7-Amino-4-hydroxy-2-naphthalenesulfonic acid, 7-Amino-4-hydroxy-2-naphthalenesulphonic acid, I-acid, J acid, 7-Amino-4-hydroxynaphthalene-2-sulphonic acid, 6-Amino-1-naphthol-3-sulphonic Acid, 6-Amino-1-naphthol-3-sulfonic Acid, 7-Amino-4-hydroxynaphthalene-2-sulfonic acid

A0370-500G A0370-25G
TCA0370-500GEA 366.56 CAD
TCA0370-500G TCA0370-025G
6-Amino-1-naphthol-3-sulfonic acid hydrate ≥95.0% (by HPLC, titration analysis)
6-Amino-1-naphthol-3-sulfonic acid hydrate

CAS Number: 87-02-5
MDL Number: MFCD00003970
Molecular Formula: C10H9NO4S
Molecular Weight: 239.25
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 7-Amino-4-hydroxy-2-naphthalenesulfonic Acid, J Acid

Formula: C₁₀H₉NO₄S
MW: 239.25 g/mol
MDL Number: MFCD00003970
CAS Number: 87-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b68471ccaa73f9ee9609686144340f4776e54c9&vid=9b68471ccaa73f9ee9609686144340f4776e54c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR