Methyl phenylglyoxylate ≥97.0%
Supplier: TCI America
Synonyms:
Methyl benzoylformate, Methyl 2-oxo-2-phenylacetate
B1033-25G
B1033-500G
B1033-100G
TCB1033-025GEA
105.86
CAD
TCB1033-025G
TCB1033-500G
TCB1033-100G
Methyl phenylglyoxylate ≥97.0%
Methyl phenylglyoxylate
CAS Number: 15206-55-0
MDL Number: MFCD00008443
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 251
Flash Point (°C): 113
Specific Gravity (20/20): 1.16
Other Product Names / Synonyms: Benzoylformic Acid Methyl Ester, Methyl Phenylglyoxylate, Phenylglyoxylic Acid Methyl Ester
Formula:
C₉H₈O₃ Boiling Pt: 251 °C Density: 1.16 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00008443 CAS Number: 15206-55-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...