Benzoin isobutyl ether ≥94.0%

Supplier: TCI America

B1015-25G B1015-500G
TCB1015-025GEA 67.94 CAD
TCB1015-025G TCB1015-500G
Benzoin isobutyl ether ≥94.0%
Benzoin isobutyl ether

CAS Number: 22499-12-3
MDL Number: MFCD00008933
Molecular Formula: C18H20O2
Molecular Weight: 268.36
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 133
Flash Point (°C): 85
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 2-Isobutoxy-2-phenylacetophenone

Formula: C₁₈H₂₀O₂
Boiling Pt: 133 °C
Density: 1.05 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00008933
CAS Number: 22499-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df01510079c2094803aea629e1585a39a8f7b5c6&vid=df01510079c2094803aea629e1585a39a8f7b5c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR