Methyl benzimidate hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

B0993-5G B0993-1G
TCB0993-5GEA 181.86 CAD
TCB0993-5G TCB0993-001G
Methyl benzimidate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Methyl benzimidate hydrochloride

[for Protein modification]
CAS Number: 5873-90-5
MDL Number: MFCD00012575
Molecular Formula: C8H9NO
Molecular Weight: 171.62
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 107
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Benzimidic Acid Methyl Ester Hydrochloride

Formula: C₈H₉NO
Melting Pt: 107 °C
MDL Number: MFCD00012575
CAS Number: 5873-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcdf155e0144dd2d424dd941d61ccde791b6fc3c&vid=fcdf155e0144dd2d424dd941d61ccde791b6fc3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR