Benzyl methyl sulfide ≥98.0%

Supplier: TCI America

Synonyms: Benzyl(methyl)sulphane, Benzyl(methyl)sulfane

B0992-25ML B0992-5ML
TCB0992-25MLEA 270.1 CAD
TCB0992-25ML TCB0992-5ML
Benzyl methyl sulfide ≥98.0%
Benzyl methyl sulfide

CAS Number: 766-92-7
MDL Number: MFCD00008563
Molecular Formula: C8H10S
Molecular Weight: 138.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 73
Specific Gravity (20/20): 1.03

Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 200 °C
Density: 1.03 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00008563
CAS Number: 766-92-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da0a6f3beb6d6e5543a52b1c9c969d183d4eb567&vid=da0a6f3beb6d6e5543a52b1c9c969d183d4eb567&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR